4-(Methoxycarbonyl)phenylboronsäurepinakolester ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

H28942.03 H28942.06
H28942.03EA 26.6 EUR
H28942.03 H28942.06
4-(Methoxycarbonyl)phenylboronsäurepinakolester ≥97%
4-(Methoxycarbonyl)phenylboronsäurepinakolester

Purity: 97% CAS No.: 171364-80-0 Molecular Formula: C14H19BO4 Formula Weight: 262.11 Melting Point: 77-81° MDL No.: MFCD02179438

Formel: C₁₄H₁₉BO₄
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179438
CAS-Nummer: 171364-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR