Kalium(3-iodphenyl)trifluoroborat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Potassium 3-iodophenyltrifluoroborate

H29009.06
H29009.06EA 95.1 EUR
H29009.06
Kalium(3-iodphenyl)trifluoroborat 96%
Potassium-3-iodophenyltrifluoroborate

Purity: 96% Molecular Formula: C6H4BF3IK Formula Weight: 309.90 Melting Point: 202-204° MDL No.: MFCD09800739

Formel: C₆H₄BIF₃K
Molecular Weight: 309,91 g/mol
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09800739
CAS-Nummer: 1189097-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR