tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tert-butyl 3-(aminomethyl)azetidine-1-carboxylate, 1-BOC-3-(aminomethyl)azetidine, tert-Butyl 3-(aminomethyl)azetidine-1-carboxylate, 3-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester
H29152.03
H29152.03EA
162
EUR
H29152.03
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate 97%
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate
Purity: 97%
CAS No.: 325775-44-8
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Refractive Index: 1.4670
MDL No.: MFCD01861762
Formel:
C₉H₁₈N₂O₂ Molecular Weight: 186,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861762 CAS-Nummer: 325775-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...