5-Pyrimidinboronsäure Hemihydrat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H29212.09
H29212.03
H29212.09EA
276
EUR
H29212.09
H29212.03
5-Pyrimidinboronsäure Hemihydrat ≥97%
5-Pyrimidinboronsäure Hemihydrat
Purity: 97%
CAS No.: 696602-91-2
Molecular Formula: C4H5BN2O2.0.5H2O
Formula Weight: 132.91 (123.91anhy)
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
C₄H₅BN₂O₂·0.5H₂O Schmelzpunkt: >300 °C |
MDL:
MFCD03002366 CAS-Nummer: 696602-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...