5-Brom-3-indolylessigsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromindol-3-essigsäure, 5-Bromindol-3-ylessigsäure
H29217.03
H29217.03EA
134
EUR
H29217.03
5-Brom-3-indolylessigsäure ≥97%
5-Brom-3-indolylessigsäure
Purity: 97%
CAS No.: 40432-84-6
Molecular Formula: C10H8BrNO2
Formula Weight: 254.09
Melting Point: 144-145° dec.
MDL No.: MFCD00005637
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Schmelzpunkt: 144…145 °C (Zers.) Dichte: 1,754 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005637 CAS-Nummer: 40432-84-6 EINECS: 254-917-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...