1-(2-Phenylethyl)-4-(phenylethynyl)benzene 97%
Lieferant: Thermo Scientific
H30058.03
H30058.03EA
104
EUR
H30058.03
1-(2-Phenylethyl)-4-(phenylethynyl)benzene 97%
1-(2-Phenylethyl)-4-(phenylethynyl)benzene
Purity: 97%
CAS No.: 906650-60-0
Molecular Formula: C22H18
Formula Weight: 282.39
Melting Point: 104-106°
MDL No.: MFCD12407075
Formel:
C₂₂H₁₈ Molecular Weight: 282,39 g/mol Schmelzpunkt: 104…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12407075 CAS-Nummer: 906650-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...