Benzophenonimin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Benzophenoneimine

H30110.06 H30110.14
H30110.06EA 73.3 EUR
H30110.06 H30110.14
Benzophenonimin 95%
Benzophenonimin

Purity: 95% CAS No.: 1013-88-3 Molecular Formula: C13H11N Formula Weight: 181.23 Refractive Index: 1.6180 Boiling Point: 151-153°/10mm MDL No.: MFCD00001760

Formel: C₁₃H₁₁N
Molecular Weight: 181.24 g/mol
Siedepunkt: 151…153 °C (10 mmHg)
Dichte: 1.08
Lagertemperatur: Raumtemperatur
MDL: MFCD00001760
CAS-Nummer: 1013-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR