ω-Brom-2-nitroacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Brommethyl-o-nitrophenylketon, 2-Brom-2'-nitroacetophenon, α-Brom-2-nitroacetophenon, 2-Nitrophenacylbromid

H30275.03 H30275.06
H30275.03EA 36.2 EUR
H30275.03 H30275.06
ω-Brom-2-nitroacetophenon ≥98%
ω-Brom-2-nitroacetophenon

Purity: 98% CAS No.: 6851-99-6 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Odour: Pungent Melting Point: 53-56° UN No.: 3261 MDL No.: MFCD00010294

Formel: C₈H₆BrNO₃
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010294
CAS-Nummer: 6851-99-6
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR