Ammoniak 7 M in Methanol

Lieferant: Thermo Fisher Scientific
Gefahr

H30382.AE H30382.AU
H30382.AEEA 108 EUR
H30382.AE H30382.AU
Ammoniak 7 M in Methanol
Ammonia

CAS No.: 7664-41-7 Molecular Formula: H3N Formula Weight: 17.03 Storage Temperature: 2°C to 8°C Flash Point: 14°(57°F) MDL No.: MFCD00011418

Formel: NH₃
Dichte: 0,779
Flammpunkt: 14 °C (57 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00011418
CAS-Nummer: 7664-41-7
EINECS: 231-635-3
UN: 3286
ADR: 3,II
Merck Index: 14,492

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR