2,5-Dibrom-4-picolin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,5-Dibrom-4-methylpyridin
H30457.06
H30457.06EA
85
EUR
H30457.06
2,5-Dibrom-4-picolin ≥97%
2,5-Dibrom-4-picolin
Purity: 97%
CAS No.: 3430-26-0
Molecular Formula: C6H5Br2N
Formula Weight: 250.92
Melting Point: 36-40°
MDL No.: MFCD00234955
Formel:
C₆H₅Br₂N Schmelzpunkt: 36…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234955 CAS-Nummer: 3430-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...