3-Brom-4-fluorbenzylalkohol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3-Bromo-4-fluorophenyl)methanol

H30604.06 H30604.14
H30604.06EA 29.5 EUR
H30604.06 H30604.14
3-Brom-4-fluorbenzylalkohol ≥97%
3-Brom-4-fluorbenzylalkohol

Purity: 97% CAS No.: 77771-03-0 Molecular Formula: C7H6BrFO Formula Weight: 205.03 Boiling Point: 80°/0.5mm MDL No.: MFCD00143093

Formel: C₇H₆BrFO
Molecular Weight: 205.03
Siedepunkt: 80 °C (0.5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00143093
CAS-Nummer: 77771-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR