Ethyl-2-amino-1,3-thiazol-4-carboxylat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-aminothiazole-4-carboxylate

H30634.09
H30634.09EA 94 EUR
H30634.09
Ethyl-2-amino-1,3-thiazol-4-carboxylat 98%
Ethyl-2-aminothiazole-4-carboxylate

Purity: 98% CAS No.: 5398-36-7 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 175-178° MDL No.: MFCD00619079

Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
Schmelzpunkt: 175…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00619079
CAS-Nummer: 5398-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR