5-Chlorpicolinsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chloro-2-pyridinecarboxylic acid, 5-Chloropyridine-2-carboxylic acid, 5-Chloro-2-picolinic acid

H30923.03
H30923.03EA 68.1 EUR
H30923.03
5-Chlorpicolinsäure 95%
5-Chlorpicolinsäure

Purity: 95% CAS No.: 86873-60-1 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: 166-171° MDL No.: MFCD04114192

Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
Schmelzpunkt: 166…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114192
CAS-Nummer: 86873-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR