2-(Ethoxycarbonyl)phenylboronsäurepinakolester ≥97%

Lieferant: Thermo Scientific

Synonyme: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

H30941.06
H30941.06EA 158 EUR
H30941.06
2-(Ethoxycarbonyl)phenylboronsäurepinakolester ≥97%
2-(Ethoxycarbonyl)phenylboronsäurepinakolester

Purity: 97% CAS No.: 269409-99-6 Molecular Formula: C15H21BO4 Formula Weight: 276.14 Melting Point: 57-62° MDL No.: MFCD03453660

Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
Schmelzpunkt: 57…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453660
CAS-Nummer: 269409-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR