2-Brom-5-formylthiazol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromothiazole-5-carboxaldehyde
H31096.03
H31096.03EA
68
EUR
H31096.03
2-Brom-5-formylthiazol 95%
2-Brom-5-formylthiazol
Purity: 95%
CAS No.: 464192-28-7
Molecular Formula: C4H2BrNOS
Formula Weight: 192.03
Melting Point: 92-96°
MDL No.: MFCD03788567
Formel:
C₄H₂BrNOS Schmelzpunkt: 92…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788567 CAS-Nummer: 464192-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...