trans-4-Aminocyclohexancarbonsäure Hydrochlorid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H31114.06
H31114.06EA
180
EUR
H31114.06
trans-4-Aminocyclohexancarbonsäure Hydrochlorid 96%
trans-4-Aminocyclohexancarbonsäure Hydrochlorid
Purity: 96%
CAS No.: 27960-59-4
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.65
Melting Point:
Formel:
C₇H₁₄ClNO₂ Molecular Weight: 179.65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844587 CAS-Nummer: 27960-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...