4-Nitro-o-toluylsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H31332.03 H31332.06
H31332.03EA 54.2 EUR
H31332.03 H31332.06
4-Nitro-o-toluylsäure ≥97%
4-Nitro-o-toluylsäure

Purity: 97% CAS No.: 1975-51-5 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odour: Odourless Melting Point: 152-155° MDL No.: MFCD00210697

Formel: C₈H₇NO₄
Molecular Weight: 181.15 g/mol
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00210697
CAS-Nummer: 1975-51-5
EINECS: 217-828-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR