4-Nitro-o-toluylsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H31332.03
H31332.06
H31332.03EA
54.2
EUR
H31332.03
H31332.06
4-Nitro-o-toluylsäure ≥97%
4-Nitro-o-toluylsäure
Purity: 97%
CAS No.: 1975-51-5
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Odour: Odourless
Melting Point: 152-155°
MDL No.: MFCD00210697
Formel:
C₈H₇NO₄ Molecular Weight: 181.15 g/mol Schmelzpunkt: 152…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210697 CAS-Nummer: 1975-51-5 EINECS: 217-828-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...