3-Chlor-p-toluidin Hydrochlorid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Chloro-4-methylaniline hydrochloride

H31342.36
H31342.36EA 0 EUR
H31342.36
3-Chlor-p-toluidin Hydrochlorid ≥97%
3-Chlor-p-toluidin Hydrochlorid

Purity: 97% CAS No.: 7745-89-3 Molecular Formula: C7H8ClN.HCl Formula Weight: 178.06 Melting Point: ca 260° subl. MDL No.: MFCD00242925

Formel: C₇H₉Cl₂N
Molecular Weight: 178,06 g/mol
Schmelzpunkt: ∼ 260 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00242925
CAS-Nummer: 7745-89-3
EINECS: 202-446-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR