4-(4-Fluorophenyl)imidazole 97%

Lieferant: Thermo Scientific
Gefahr

H31503.06
H31503.06EA 78.9 EUR
H31503.06
4-(4-Fluorophenyl)imidazole 97%
4-(4-Fluorophenyl)imidazole

Purity: 97% CAS No.: 65020-70-4 Molecular Formula: C9H7FN2 Formula Weight: 162.17 Melting Point: 127-131° MDL No.: MFCD00297041

Formel: C₉H₇FN₂
Molecular Weight: 162,17 g/mol
Schmelzpunkt: 127…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00297041
CAS-Nummer: 65020-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR