Chlorwasserstoff 5->6 M in 2-Propanol, rein

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chlorwasserstoff

H31519.AE H31519.AP
H31519.AEEA 63.2 EUR
H31519.AE H31519.AP
Chlorwasserstoff 5->6 M in 2-Propanol, rein
Chlorwasserstoff

CAS No.: 7647-01-0 Molecular Formula: ClH Formula Weight: 36.46 Storage Temperature: 2°C to 8°C Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.909 Sensitivity: Hygroscopic Appearance: Colourless Odour: Alcohol-like UN No.: 2924 MDL No.: MFCD00011324

Formel: HCl
Molecular Weight: 36.46 g/mol
Dichte: 0.909
Flammpunkt: 11 °C (52 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR