Ethyl-2-chlor-4-oxazolcarboxylat 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-chlorooxazole-4-carboxylate

H31521.03
H31521.03EA 91 EUR
H31521.03
Ethyl-2-chlor-4-oxazolcarboxylat 95%
Ethyl-2-chlor-4-oxazolcarboxylat

Purity: 95% CAS No.: 46081-18-9 Molecular Formula: C6H6ClNO3 Formula Weight: 175.57 Melting Point: 97-98° MDL No.: MFCD06660120

Formel: C₆H₆ClNO₃
Molecular Weight: 175,57 g/mol
Schmelzpunkt: 97…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660120
CAS-Nummer: 460081-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR