2-Brom-5-methylbenzotrifluorid ≥98%

Lieferant: Thermo Scientific
Achtung

H31523.03
H31523.03EA 79.1 EUR
H31523.03
2-Brom-5-methylbenzotrifluorid ≥98%
2-Brom-5-methylbenzotrifluorid

Purity: 98% CAS No.: 261952-20-9 Molecular Formula: C8H6BrF3 Formula Weight: 239.04 MDL No.: MFCD01631586

Formel: C₈H₆BrF₃
Molecular Weight: 239,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631586
CAS-Nummer: 261952-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR