2-Brom-3'-(trifluormethyl)acetophenon 98

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-(3-(trifluoromethyl)phenyl)ethanone

H31526.03 H31526.06 H31526.09
H31526.03EA 88.4 EUR
H31526.03 H31526.06 H31526.09
2-Brom-3'-(trifluormethyl)acetophenon 98
2-Brom-3'-(trifluormethyl)acetophenon

Purity: 98% CAS No.: 2003-10-3 Molecular Formula: C9H6BrF3O Formula Weight: 267.04 Physical Form: Fused solid Appearance: Pale yellow Melting Point: 20-23° Boiling Point: 69-72° MDL No.: MFCD03094283

Formel: C₉H₆BrF₃O
Molecular Weight: 267.04 g/mol
Siedepunkt: 69…72 °C
Schmelzpunkt: 20…23 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094283
CAS-Nummer: 2003-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR