2,3-Dimethyl-4-fluoranilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Amino-6-fluor-o-xylol, 4-Fluor-2,3-xylidin

H31560.MD H31560.03
H31560.MDEA 70.6 EUR
H31560.MD H31560.03
2,3-Dimethyl-4-fluoranilin ≥97%
2,3-Dimethyl-4-fluoranilin

Purity: 97% CAS No.: 1737-68-4 Molecular Formula: C8H10FN Formula Weight: 139.17 Melting Point: 42-46° UN No.: 2811 MDL No.: MFCD04972870

Formel: C₈H₁₀FN
Schmelzpunkt: 42…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972870
CAS-Nummer: 1737-68-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR