4-(4-Bromophenyl)imidazole 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H31581.06
H31581.06EA
72.4
EUR
H31581.06
4-(4-Bromophenyl)imidazole 97%
4-(4-Bromophenyl)imidazole
Purity: 97%
CAS No.: 13569-96-5
Molecular Formula: C9H7BrN2
Formula Weight: 223.07
Melting Point: 138-142°
MDL No.: MFCD03312935
Formel:
C₉H₇BrN₂ Molecular Weight: 223,07 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03312935 CAS-Nummer: 13569-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...