3,5-Difluor-4-hydroxybenzonitril ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H31620.MD
H31620.03
H31620.MDEA
72.3
EUR
H31620.MD
H31620.03
3,5-Difluor-4-hydroxybenzonitril ≥97%
3,5-Difluor-4-hydroxybenzonitril
Purity: 97%
CAS No.: 2967-54-6
Molecular Formula: C7H3F2NO
Formula Weight: 155.10
Melting Point: 200-208°
UN No.: 3439
MDL No.: MFCD09033181
Formel:
C₇H₃F₂NO Schmelzpunkt: 200…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033181 CAS-Nummer: 2967-54-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...