3,5-Difluor-4-hydroxybenzonitril ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H31620.MD H31620.03
H31620.MDEA 72.3 EUR
H31620.MD H31620.03
3,5-Difluor-4-hydroxybenzonitril ≥97%
3,5-Difluor-4-hydroxybenzonitril

Purity: 97% CAS No.: 2967-54-6 Molecular Formula: C7H3F2NO Formula Weight: 155.10 Melting Point: 200-208° UN No.: 3439 MDL No.: MFCD09033181

Formel: C₇H₃F₂NO
Schmelzpunkt: 200…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09033181
CAS-Nummer: 2967-54-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR