Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate

H31668.03
H31668.03EA 181 EUR
H31668.03
Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate 97%
Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate

Purity: 97% CAS No.: 59604-96-5 Molecular Formula: C12H14O3 Formula Weight: 206.24 Melting Point: 40-43° MDL No.: MFCD09258894

Formel: C₁₂H₁₄O₃
Molecular Weight: 206,24 g/mol
Schmelzpunkt: 40…43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258894
CAS-Nummer: 59604-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR