5-Nitro-3-picolin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Methyl-5-nitropyridine
H31683.06
H31683.06EA
124
EUR
H31683.06
5-Nitro-3-picolin ≥97%
5-Nitro-3-picolin
Purity: 97%
CAS No.: 6960-20-9
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: 94-98°
UN No.: 2811
MDL No.: MFCD04114214
Formel:
C₆H₆N₂O₂ Molecular Weight: 138.13 g/mol Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114214 CAS-Nummer: 6960-20-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...