4'-Hydroxy-3'-(trifluormethyl)acetophenon 95%
Lieferant: Thermo Scientific
|
Achtung
|
H31691.03
H31691.03EA
81.4
EUR
H31691.03
4'-Hydroxy-3'-(trifluormethyl)acetophenon 95%
4'-Hydroxy-3'-(trifluormethyl)acetophenon
Purity: 95%
CAS No.: 149105-11-3
Molecular Formula: C9H7F3O2
Formula Weight: 204.15
Melting Point: 174-177°
MDL No.: MFCD01091005
Formel:
C₉H₇F₃O₂ Schmelzpunkt: 174…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01091005 CAS-Nummer: 149105-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...