2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

H31692.MD
H31692.MDEA 264 EUR
H31692.MD
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester

Purity: 95% CAS No.: 1315281-29-8 Molecular Formula: C10H16BClN2O2 Formula Weight: 242.52 Storage Temperature: 2°C to 8°C Melting Point: 131-135° MDL No.: MFCD11100959

Formel: C₁₀H₁₆BClN₂O₂
Molecular Weight: 242.52 g/mol
Schmelzpunkt: 131…135 °C
MDL: MFCD11100959
CAS-Nummer: 1315281-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR