3-Fluor-2-hydroxypyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H31705.MD H31705.03 H31705.06
H31705.MDEA 95 EUR
H31705.MD H31705.03 H31705.06
3-Fluor-2-hydroxypyridin ≥97%
3-Fluor-2-hydroxypyridin

Purity: 97% CAS No.: 1547-29-1 Molecular Formula: C5H4FNO Formula Weight: 113.09 Odour: Odourless Melting Point: 157-161° MDL No.: MFCD03095042

Formel: C₅H₄FNO
Molecular Weight: 113.09 g/mol
Schmelzpunkt: 157…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095042
CAS-Nummer: 1547-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR