2,3-Dibromisonicotinsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,3-Dibromopyridine-4-carboxylic acid

H31708.03
H31708.03EA 130 EUR
H31708.03
2,3-Dibromisonicotinsäure ≥97%
2,3-Dibromisonicotinsäure

Purity: 97% CAS No.: 1020056-98-7 Molecular Formula: C6H3Br2NO2 Formula Weight: 280.90 MDL No.: MFCD09997708

Formel: C₆H₃Br₂NO₂
Molecular Weight: 280,90 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997708
CAS-Nummer: 1020056-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR