4-Fluor-2-(trifluormethyl)phenol 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Fluoro-2-hydroxybenzotrifluoride

H31716.03
H31716.03EA 116 EUR
H31716.03
4-Fluor-2-(trifluormethyl)phenol 95%
4-Fluor-2-(trifluormethyl)phenol

Purity: 95% CAS No.: 130047-19-7 Molecular Formula: C7H4F4O Formula Weight: 180.10 Melting Point: 50-54° UN No.: 3261 MDL No.: MFCD00061290

Formel: C₇H₄F₄O
Molecular Weight: 180,1 g/mol
Schmelzpunkt: 50…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061290
CAS-Nummer: 130047-19-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR