2-Bromo-3-fluoropropionic acid 96%
Lieferant: Thermo Scientific
|
Gefahr
|
H31718.03
H31718.03EA
157
EUR
H31718.03
2-Bromo-3-fluoropropionic acid 96%
2-Bromo-3-fluoropropionic acid
Purity: 96%
Molecular Formula: C3H4BrFO2
Formula Weight: 170.97
Boiling Point: 62-63°/0.2mm
UN No.: UN3265
MDL No.: MFCD09800640
Formel:
C₃H₄BrFO₂ Siedepunkt: 62…63 °C (0,2 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800640 CAS-Nummer: 39621-37-9 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...