4-Brom-2-(trifluormethoxy)phenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H31726.06
H31726.06EA 166 EUR
H31726.06
4-Brom-2-(trifluormethoxy)phenol ≥98%
4-Brom-2-(trifluormethoxy)phenol

Purity: 98% CAS No.: 690264-39-2 Molecular Formula: C7H4BrF3O2 Formula Weight: 257.01 Melting Point: 51-55° MDL No.: MFCD09037798

Formel: C₇H₄BrF₃O₂
Schmelzpunkt: 51…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09037798
CAS-Nummer: 690264-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR