5-Brom-2-(trifluormethoxy)benzaldehyd 98%

Lieferant: Thermo Scientific
Achtung

H31751.09
H31751.09EA 161 EUR
H31751.09
5-Brom-2-(trifluormethoxy)benzaldehyd 98%
5-Brom-2-(trifluormethoxy)benzaldehyd

Purity: 98% CAS No.: 923281-52-1 Molecular Formula: C8H4BrF3O2 Formula Weight: 269.01 MDL No.: MFCD03701029

Formel: C₈H₄BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03701029
CAS-Nummer: 923281-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR