4-Pyrimidinemethanol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Hydroxymethyl)pyrimidine

H31754.MD
H31754.MDEA 170 EUR
H31754.MD
4-Pyrimidinemethanol 98%
4-Pyrimidinemethanol

Purity: 98% CAS No.: 33581-98-5 Molecular Formula: C5H6N2O Formula Weight: 110.12 Melting Point: 75-78° MDL No.: MFCD03789625

Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 75…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03789625
CAS-Nummer: 33581-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR