3,4-Pyridinedicarboximide

Lieferant: Thermo Fisher Scientific
Achtung

H31758.06 H31758.14 H31758.22
H31758.06EA 42.6 EUR
H31758.06 H31758.14 H31758.22
3,4-Pyridinedicarboximide
3,4-Pyridinedicarboximide

CAS No.: 4664-01-1 Molecular Formula: C7H4N2O2 Formula Weight: 148.12 Melting Point: 230-232° MDL No.: MFCD00013439

Formel: C₇H₄N₂O₂
Schmelzpunkt: 230…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013439
CAS-Nummer: 4664-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR