Ethyl-2-amino-1,3-thiazol-5-carboxylat 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl 2-aminothiazole-5-carboxylate
H31769.03
H31769.06
H31769.03EA
37.2
EUR
H31769.03
H31769.06
Ethyl-2-amino-1,3-thiazol-5-carboxylat 97%
Ethyl-2-aminothiazole-5-carboxylate
Purity: 97%
CAS No.: 32955-21-8
Molecular Formula: C6H8N2O2S
Formula Weight: 172.21
Storage Temperature: 2°C to 8°C
Melting Point: 158-160°
MDL No.: MFCD00602139
Formel:
C₆H₈N₂O₂S Molecular Weight: 172.21 g/mol Schmelzpunkt: 158…160 °C |
MDL:
MFCD00602139 CAS-Nummer: 32955-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...