(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl 97%
Lieferant: Thermo Fisher Scientific
H31791.03
H31791.03EA
253
EUR
H31791.03
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl 97%
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Purity: 97%
CAS No.: 173831-50-0
Molecular Formula: C24H22O4
Formula Weight: 374.44
Melting Point: 101-105°
Optical Rotation: +94.5°(C=1 in THF)
MDL No.: MFCD03788935
Formel:
C₂₄H₂₂O₄ Schmelzpunkt: 101…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788935 CAS-Nummer: 173831-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...