4-(2-Thiazolyl)phenol 97%

Lieferant: Thermo Fisher Scientific
Achtung

H31827.03 H31827.09
H31827.03EA 25.4 EUR
H31827.03 H31827.09
4-(2-Thiazolyl)phenol 97%
4-(2-Thiazolyl)phenol

Purity: 97% CAS No.: 81015-49-8 Molecular Formula: C9H7NOS Formula Weight: 177.28 Melting Point: 162-165° MDL No.: MFCD07368615

Formel: C₉H₇NOS
Schmelzpunkt: 162…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368615
CAS-Nummer: 81015-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR