2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H31846.MD
H31846.03
H31846.MDEA
119
EUR
H31846.MD
H31846.03
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole
Purity: 95%
Molecular Formula: C8H11IN2O
Formula Weight: 278.09
MDL No.: MFCD11100955
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...