Methyl-1,2,3,4-Tetra-O-Acetyl-Beta-D-Glucuronat 98%

Lieferant: Thermo Scientific

Synonyme: Methyl 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronate, (2S,3R,4S,5S,6S)-6-(Methoxycarbonyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4-Tetra-O-acetyl-β-D-glucuronic acid Methyl Ester, 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronic acid Methyl Ester, Methyl tetra-O-acetyl-β-D-glucopyranuronate

H31888.03
H31888.03EA 190 EUR
H31888.03
Methyl-1,2,3,4-Tetra-O-Acetyl-Beta-D-Glucuronat 98%
Methyl-1,2,3,4-tetra-O-acetyl-beta-D-glucuronate

Purity: 98% CAS No.: 7355-18-2 Molecular Formula: C15H20O11 Formula Weight: 376.32 Odour: Odourless Melting Point: 177-181° Optical Rotation: +8.5° (c=1 in chloroform) MDL No.: MFCD00069834

Formel: C₁₅H₂₀O₁₁
Molecular Weight: 376,31 g/mol
Schmelzpunkt: 177…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069834
CAS-Nummer: 7355-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR