2-Fluor-α-methylbenzylalkohol 95%

Lieferant: Thermo Scientific

H31900.09
H31900.09EA 76.5 EUR
H31900.09
2-Fluor-α-methylbenzylalkohol 95%
2-Fluor-α-methylbenzylalkohol

Purity: 95% CAS No.: 445-26-1 Molecular Formula: C8H9FO Formula Weight: 140.16 MDL No.: MFCD00014401

Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014401
CAS-Nummer: 445-26-1
EINECS: 207-155-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR