2-Amino-3-fluorpyridin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H31970.03
H31970.03EA
109
EUR
H31970.03
2-Amino-3-fluorpyridin 97%
2-Amino-3-fluorpyridin
Purity: 97%
CAS No.: 21717-95-3
Molecular Formula: C5H5FN2
Formula Weight: 112.11
Odour: Characteristic
Melting Point: 39-43°
UN No.: 2811
MDL No.: MFCD04114135
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Schmelzpunkt: 39…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114135 CAS-Nummer: 21717-95-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...