3-Fluor-4-nitrobenzolsulfonylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Fluor-4-nitrobenzol-1-sulfonylchlorid, 3-Fluor-4-nitrobenzolsulfochlorid, 3-Fluor-4-nitrobenzolsulfonsäurechlorid

H31979.03
H31979.03EA 83 EUR
H31979.03
3-Fluor-4-nitrobenzolsulfonylchlorid ≥98%
3-Fluor-4-nitrobenzolsulfonylchlorid

Purity: 98% CAS No.: 86156-93-6 Molecular Formula: C6H3ClFNO4S Formula Weight: 239.61 Melting Point: 44-48° UN No.: 3261 MDL No.: MFCD03412240

Formel: C₆H₃ClFNO₄S
Molecular Weight: 239,61 g/mol
Schmelzpunkt: 44…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412240
CAS-Nummer: 86156-93-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR