2-Brom-3-fluor-5-(trifluormethyl)pyridin ≥97%

Lieferant: Thermo Scientific
Achtung

H31993.03
H31993.03EA 194 EUR
H31993.03
2-Brom-3-fluor-5-(trifluormethyl)pyridin ≥97%
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 89402-29-9 Molecular Formula: C6H2BrF4N Formula Weight: 243.99 Refractive Index: 1.4640

Formel: C₆H₂BrF₄N
Molecular Weight: 243,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699118
CAS-Nummer: 89402-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR