Methyl-3-fluor-4-methoxybenzoat 95%

Lieferant: Thermo Scientific

Synonyme: 3-Fluor-4-methoxybenzoesäuremethylester

H31994.09
H31994.09EA 104 EUR
H31994.09
Methyl-3-fluor-4-methoxybenzoat 95%
Methyl-3-fluor-4-methoxybenzoat

Purity: 95% CAS No.: 369-30-2 Molecular Formula: C9H9FO3 Formula Weight: 184.17 Melting Point: 69-73° MDL No.: MFCD00215839

Formel: C₉H₉FO₃
Molecular Weight: 184,17 g/mol
Schmelzpunkt: 69…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215839
CAS-Nummer: 369-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR